missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Tri-n-butylstannyl)oxazole, 97%

Manufacturer:  Alfa Aesar™ H5587003

 View more versions of this product

Catalog No. AAH5587003



Description

Description

Applications
2-(Tri-n-butylstannyl)oxazole act as a synthetic building block, which is utilized in Stille coupling

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

2-(Tri-n-butylstannyl)oxazole
>110°C (230°F)
108°C to 110°C (0.2mmHg)
1g
C15H29NOSn
UN2788
Insoluble in water.
CCCC[Sn](CCCC)(CCCC)C1=NC=CO1
358.113
358.11
97%
1.71 g/mL
1.493
Colorless
145214-05-7
MFCD06798076
2-tri-n-butylstannyl oxazole, 2-tributylstannanyl-oxazole, 2-tributylstannanyloxazole, 2-tributylstannyl oxazole, 2-tributylstannyl-1,3-oxazole, 2-tributylstannyl-oxazole, 2-tributylstannyloxazole, 2tributylstannyloxazole, acmc-20aom1
YOWGRWHKDCHINP-UHFFFAOYSA-N
tributyl(1,3-oxazol-2-yl)stannane
15486183
Liquid

Chemical Identifiers

145214-05-7
358.113
YOWGRWHKDCHINP-UHFFFAOYSA-N
15486183
CCCC[Sn](CCCC)(CCCC)C1=NC=CO1
C15H29NOSn
MFCD06798076
2-tri-n-butylstannyl oxazole, 2-tributylstannanyl-oxazole, 2-tributylstannanyloxazole, 2-tributylstannyl oxazole, 2-tributylstannyl-1,3-oxazole, 2-tributylstannyl-oxazole, 2-tributylstannyloxazole, 2tributylstannyloxazole, acmc-20aom1
tributyl(1,3-oxazol-2-yl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents