missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Tri-n-butylstannyl)furan, 97%

Manufacturer:  Alfa Aesar™ H55259AC

 View more versions of this product

Catalog No. AAH55259AC



Description

Description

Applications
It is an agrochemical, organic and pharmaceutical intermediates.

Notes
Store at room temperature. Light sensitive.
Specifications

Specifications

2-(Tri-n-butylstannyl)furan
1.4940
Colorless to Yellow
118486-94-5
MFCD00192512
2-furyltributyltin
CCCC[Sn](CCCC)(CCCC)C1=CC=CO1
357.125
357.1
97%
1.134g/mL
90°C (0.1mmHg)
25mL
C16H30OSn
UN2788
SANWDQJIWZEKOD-UHFFFAOYSA-N
tributyl(furan-2-yl)stannane
4393688
Liquid

Chemical Identifiers

118486-94-5
357.125
SANWDQJIWZEKOD-UHFFFAOYSA-N
4393688
CCCC[Sn](CCCC)(CCCC)C1=CC=CO1
C16H30OSn
MFCD00192512
2-furyltributyltin
tributyl(furan-2-yl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

Documents