missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[trans-4-(Boc-amino)cyclohexyl]ethylamine, 97%, Alfa Aesar™

$382.45 - $1,221.36

Chemical Identifiers

CAS 1212272-05-3
Molecular Formula C13H26N2O2
Molecular Weight (g/mol) 242.363
MDL Number MFCD03844600
InChI Key SSZKBGFWNFAXNC-UHFFFAOYSA-N
Synonym tert-butyl 4-2-aminoethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino-cyclohexyl ethylamine, tert-butyl cis-4-2-aminoethyl cyclohexylcarbamate, carbamicacid, cis-4-2-aminoethyl cyclohexyl-, 1,1-dimethylethyl ester 9ci, tert-butyl n-4-2-aminoethyl cyclohexyl carbamate, tert-butyl n-4-2-azanylethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino cyclohexyl ethylamine, tert-butyl 4-2-aminoethyl cyclohexyl carbamate, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate
PubChem CID 24720893
IUPAC Name tert-butyl N-[4-(2-aminoethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62326MD View Documents Alfa Aesar
H62326MD
250mg Each for $382.45
 
AAH6232603 View Documents Alfa Aesar
H6232603
1g Each for $1,221.36
 
Description

Description

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1212272-05-3
242.363
SSZKBGFWNFAXNC-UHFFFAOYSA-N
24720893
CC(C)(C)OC(=O)NC1CCC(CC1)CCN
C13H26N2O2
MFCD03844600
tert-butyl 4-2-aminoethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino-cyclohexyl ethylamine, tert-butyl cis-4-2-aminoethyl cyclohexylcarbamate, carbamicacid, cis-4-2-aminoethyl cyclohexyl-, 1,1-dimethylethyl ester 9ci, tert-butyl n-4-2-aminoethyl cyclohexyl carbamate, tert-butyl n-4-2-azanylethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino cyclohexyl ethylamine, tert-butyl 4-2-aminoethyl cyclohexyl carbamate, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate
tert-butyl N-[4-(2-aminoethyl)cyclohexyl]carbamate

Specifications

250mg
C13H26N2O2
tert-butyl 4-2-aminoethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino-cyclohexyl ethylamine, tert-butyl cis-4-2-aminoethyl cyclohexylcarbamate, carbamicacid, cis-4-2-aminoethyl cyclohexyl-, 1,1-dimethylethyl ester 9ci, tert-butyl n-4-2-aminoethyl cyclohexyl carbamate, tert-butyl n-4-2-azanylethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino cyclohexyl ethylamine, tert-butyl 4-2-aminoethyl cyclohexyl carbamate, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate
CC(C)(C)OC(=O)NC1CCC(CC1)CCN
242.363
242.35
2-[trans-4-(Boc-amino)cyclohexyl]ethylamine
1212272-05-3
MFCD03844600
SSZKBGFWNFAXNC-UHFFFAOYSA-N
tert-butyl N-[4-(2-aminoethyl)cyclohexyl]carbamate
24720893
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning