Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone, 95%

Chemical Identifiers

Chemical Name or Material 2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone
CAS 315-08-2
Formula Weight 277.28
MDL Number MFCD00126176
Synonym 3-[3-(Trifluoromethyl)phenyl]rhodanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6344703

  SDS
alfa aesar™
H6344703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6344706

  SDS
alfa aesar™
H6344706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63447MD

  SDS
alfa aesar™
H63447MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 277.28
Chemical Name or Material 2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone
Physical Form Solid
Assay 95%
MDL Number MFCD00126176
CAS 315-08-2
Synonym 3-[3-(Trifluoromethyl)phenyl]rhodanine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Off-white
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.