Learn More
2-Thioxanthine, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2160603
Description
2-Thioxanthine used to inhibit MPO (Myeloperoxidase).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Thioxanthine used to inhibit MPO (Myeloperoxidase).
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
2-Thioxanthine | |
>300°C | |
MFCD00031505 | |
2-thioxanthine, 6-hydroxy-2-mercaptopurine, 2-mercapto-9h-purin-6-ol, 2-thioxanthene, 2-mercapto-6-hydroxypurine, 2-thio-6-oxypurine, 6-hydroxypurine-2-thiol, 2-thioxo-2,3-dihydro-1h-purin-6 7h-one, xanthine, 2-thio, 6h-purin-6-one, 1,2,3,7-tetrahydro-2-thioxo | |
XNHFAGRBSMMFKL-UHFFFAOYSA-N | |
2-sulfanylidene-3,7-dihydropurin-6-one | |
1268185 | |
≥98% |
2487-40-3 | |
C5H4N4OS | |
1 g | |
Slightly soluble in water. | |
C1=NC2=C(N1)C(=O)NC(=S)N2 | |
168.174 | |
168.17 |
Chemical Identifiers
2487-40-3 | |
168.174 | |
XNHFAGRBSMMFKL-UHFFFAOYSA-N | |
1268185 | |
C1=NC2=C(N1)C(=O)NC(=S)N2 |
C5H4N4OS | |
MFCD00031505 | |
2-thioxanthine, 6-hydroxy-2-mercaptopurine, 2-mercapto-9h-purin-6-ol, 2-thioxanthene, 2-mercapto-6-hydroxypurine, 2-thio-6-oxypurine, 6-hydroxypurine-2-thiol, 2-thioxo-2,3-dihydro-1h-purin-6 7h-one, xanthine, 2-thio, 6h-purin-6-one, 1,2,3,7-tetrahydro-2-thioxo | |
2-sulfanylidene-3,7-dihydropurin-6-one |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 219-636-7
RTECSNumber : UP0825000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only