Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Thiouracil, 98%

$92.36 - $326.97

Chemical Identifiers

Chemical Name or Material 2-Thiouracil
CAS 141-90-2
Molecular Formula C4H4N2OS
Formula Weight 128.15
MDL Number MFCD00006039
Beilstein 112227
Merck Index 14,9366
Synonym 4-Hydroxy-2-mercaptopyrimidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1811922

  SDS
alfa aesar™
A1811922
100g Each for $92.36

AAA1811936

  SDS
alfa aesar™
A1811936
500g Each for $326.97
Description & Specifications

Specifications

Formula Weight 128.15
Chemical Name or Material 2-Thiouracil
TSCA TSCA
Assay 98%
Molecular Formula C4H4N2OS
MDL Number MFCD00006039
CAS 141-90-2
Beilstein 112227
Merck Index 14,9366
Synonym 4-Hydroxy-2-mercaptopyrimidine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point >300°C
Quantity 100g
RTECS Number YR1575000
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.