missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Thiophenemethanol, 97%, Thermo Scientific Chemicals
$104.58 - $260.73
Chemical Identifiers
CAS | 636-72-6 |
---|---|
Molecular Formula | C5H6OS |
Molecular Weight (g/mol) | 114.16 |
MDL Number | MFCD00005454 |
InChI Key | ZPHGMBGIFODUMF-UHFFFAOYSA-N |
Synonym | 2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol |
PubChem CID | 69467 |
IUPAC Name | thiophen-2-ylmethanol |
SMILES | OCC1=CC=CS1 |
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
AAA1576814
|
Thermo Scientific Chemicals
A1576814 |
25 g |
Each for $104.58
|
|
|||||
AAA1576822
|
Thermo Scientific Chemicals
A1576822 |
100 g |
Each for $260.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
636-72-6 | |
114.16 | |
ZPHGMBGIFODUMF-UHFFFAOYSA-N | |
69467 | |
OCC1=CC=CS1 |
C5H6OS | |
MFCD00005454 | |
2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol | |
thiophen-2-ylmethanol |
Specifications
636-72-6 | |
1.206 | |
>110°C (230°F) | |
1.565 | |
25 g | |
Air Sensitive | |
ZPHGMBGIFODUMF-UHFFFAOYSA-N | |
thiophen-2-ylmethanol | |
69467 | |
97% |
0°C | |
206°C to 207°C | |
C5H6OS | |
MFCD00005454 | |
383595 | |
2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol | |
OCC1=CC=CS1 | |
114.16 | |
114.17 | |
2-Thiophenemethanol |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H312-H315-H319-H332-H335
EINECSNumber : 211-264-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only