missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Thiophenemethanethiol 95.0+%, TCI America™
Supplier: TCI America T26135G
Specifications
2-Thiophenemethanethiol | |
Yellow | |
C5H6S2 | |
5 g | |
GCZQHDFWKVMZOE-UHFFFAOYSA-N | |
thiophen-2-ylmethanethiol | |
80408 | |
≥95.0% (GC) |
6258-63-5 | |
86°C | |
MFCD00107135 | |
2-thiophenemethanethiol, thenyl mercaptan, 2-thenylmercaptan, 2-thienylmethanethiol, thiophene-2-methanethiol, 2-thenylthiol, thiophen-2-yl-methanethiol, unii-9jml279xs6, 2-thenyl mercaptan, 2-mercaptomethyl thiophene | |
C1=CSC(=C1)CS | |
130.223 | |
130.22 | |
Liquid |
Chemical Identifiers
6258-63-5 | |
130.223 | |
GCZQHDFWKVMZOE-UHFFFAOYSA-N | |
80408 | |
C1=CSC(=C1)CS |
C5H6S2 | |
MFCD00107135 | |
2-thiophenemethanethiol, thenyl mercaptan, 2-thenylmercaptan, 2-thienylmethanethiol, thiophene-2-methanethiol, 2-thenylthiol, thiophen-2-yl-methanethiol, unii-9jml279xs6, 2-thenyl mercaptan, 2-mercaptomethyl thiophene | |
thiophen-2-ylmethanethiol |
Safety and Handling
TSCA : No