missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Thiophenecarboxylic acid hydrazide, 97%, ACROS Organics™

Chemical Identifiers

CAS 2361-27-5
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.18
MDL Number MFCD00005435
InChI Key SOGBOGBTIKMGFS-UHFFFAOYSA-N
Synonym 2-thenoylhydrazine, 2-thienylcarboxylic acid hydrazide, 2-thiophene carboxylic hydrazide, 2-thiophenecarbohydrazide, 2-thiophenecarbohydrazonic acid, 2-thiophenecarboxylic acid hydrazide, 2-thiophenecarboxylic acid, hydrazide, 2-thiophenecarboxylic hydrazide, thiophene-2-carboxylic acid hydrazide, thiophene-2-carboxylic hydrazide
PubChem CID 73098
IUPAC Name thiophene-2-carbohydrazide
SMILES C1=CSC(=C1)C(=O)NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC138850050
SDS View Documents
Thermo Fisher Scientific
138850050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC138850250
SDS View Documents
Thermo Fisher Scientific
138850250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

2361-27-5
142.18
SOGBOGBTIKMGFS-UHFFFAOYSA-N
73098
C1=CSC(=C1)C(=O)NN
C5H6N2OS
MFCD00005435
2-thenoylhydrazine, 2-thienylcarboxylic acid hydrazide, 2-thiophene carboxylic hydrazide, 2-thiophenecarbohydrazide, 2-thiophenecarbohydrazonic acid, 2-thiophenecarboxylic acid hydrazide, 2-thiophenecarboxylic acid, hydrazide, 2-thiophenecarboxylic hydrazide, thiophene-2-carboxylic acid hydrazide, thiophene-2-carboxylic hydrazide
thiophene-2-carbohydrazide

Specifications

Authentic
Yellow to Beige
5g
96.0
96% min. (GC)
MFCD00005435
2-thenoylhydrazine, 2-thienylcarboxylic acid hydrazide, 2-thiophene carboxylic hydrazide, 2-thiophenecarbohydrazide, 2-thiophenecarbohydrazonic acid, 2-thiophenecarboxylic acid hydrazide, 2-thiophenecarboxylic acid, hydrazide, 2-thiophenecarboxylic hydrazide, thiophene-2-carboxylic acid hydrazide, thiophene-2-carboxylic hydrazide
C1=CSC(=C1)C(=O)NN
142.18
142.18
97%
Glass bottle
136.0°C to 139.0°C
2361-27-5
100.0
C5H6N2OS
18, 291
SOGBOGBTIKMGFS-UHFFFAOYSA-N
thiophene-2-carbohydrazide
73098
Crystalline Powder
2-Thiophenecarboxylic acid hydrazide, 97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 219-107-0

TSCA : TSCA

SDS