Learn More
2-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 138840050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Thiophenecarboxylic acid | |
98.5 | |
126.0°C to 130.0°C | |
98.5% min. (GC) | |
C5H4O2S | |
5 g | |
15,9507 | |
Solubility in water: 80g/L (20°C). Other solubilities: soluble in alcohol and ether, moderately soluble in chcl3, slightly soluble in petroleum ether | |
OC(=O)C1=CC=CS1 | |
128.15 | |
CHEBI:71241 | |
99% |
527-72-0 | |
100.0 | |
260.0°C | |
Glass bottle | |
MFCD00005437 | |
18,289 | |
2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid | |
QERYCTSHXKAMIS-UHFFFAOYSA-N | |
thiophene-2-carboxylic acid | |
10700 | |
128.15 |
Chemical Identifiers
527-72-0 | |
128.15 | |
QERYCTSHXKAMIS-UHFFFAOYSA-N | |
10700 | |
thiophene-2-carboxylic acid |
C5H4O2S | |
MFCD00005437 | |
2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid | |
CHEBI:71241 | |
OC(=O)C1=CC=CS1 |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 208-423-4
RUO – Research Use Only