missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Thiophenecarboxamide, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC305420250

 View more versions of this product

Catalog No. AC305420250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Thiophenecarboxamide
98.5
Authentic
≥98.5%
C5H5NOS
127.17
White to grayish
25g
5813-89-8
100.0
Crystalline Powder or Crystals
99%
Glass bottle
MFCD00010425
179° to 182°C

Chemical Identifiers

5813-89-8
100.0
C5H5NOS
MFCD00010425
≥98.5%
98.5
99%
127.17
2-Thiophenecarboxamide

Safety and Handling

SDS & Safety Documents

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning