missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Thiophenecarbonitrile, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC298052500

 View more versions of this product

Catalog No. AC298052500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Thiophenecarbonitrile
1003-31-2
100.0
Liquid
97%
Glass bottle
MFCD00005416
2-Cyanothiophene
Clear colorless to yellow
97%
96.0
Authentic
≥96.0%
C5H3NS
109.15
1.5620 to 1.5640 (20°C, 589nm)
1.1700g/mL
250mL

Chemical Identifiers

1003-31-2
100.0
C5H3NS
MFCD00005416
2-Thiophenecarbonitrile, 97%
96.0
97%
109.15
2-Cyanothiophene
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-706-0

TSCA : TSCA