Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Thiopheneacetyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2-Thiopheneacetyl chloride
CAS 39098-97-0
Molecular Formula C6H5ClOS
Formula Weight 160.62
MDL Number MFCD00005456
UN Number UN3265
Beilstein 114547
Synonym 2-Thienylacetyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0932306

  SDS
alfa aesar™
L0932306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0932314

  SDS
alfa aesar™
L0932314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.62
CAS 39098-97-0
Chemical Name or Material 2-Thiopheneacetyl chloride
TSCA TSCA
Assay 98%
Molecular Formula C6H5ClOS
MDL Number MFCD00005456
UN Number UN3265
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Boiling Point 131° to 132°C (90mmHg)
Quantity 5g
Beilstein 114547
Synonym 2-Thienylacetyl chloride
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point 101°C (213°F)

Applications
2-Thiopheneacetyl chloride was used in the synthesis of (S)-ethyl-1-(2-thiopheneacetyl)-3-piperidinecarboxylate, nipecotate-containing immunopotentiator, 5-fluorouracil-cephalosporin prodrug, series of new N,N′-di(thiopheneacetyl)diamines derivatives and 6-β-(thiophen-2′-yl)acetamidomorphine. Also used in the synthesis of Nitrocefin.

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.