Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Thiopheneacetonitrile, 96%

Chemical Identifiers

Chemical Name or Material 2-Thiopheneacetonitrile
CAS 20893-30-5
Molecular Formula C6H5NS
Formula Weight 123.18
MDL Number MFCD00005453
UN Number UN3276
Beilstein 106960
Synonym 2-Thienylacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1504722

  SDS
alfa aesar™
A1504722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1504736

  SDS
alfa aesar™
A1504736
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 123.18
Chemical Name or Material 2-Thiopheneacetonitrile
TSCA TSCA
Assay 96%
Molecular Formula C6H5NS
MDL Number MFCD00005453
CAS 20893-30-5
UN Number UN3276
Beilstein 106960
Synonym 2-Thienylacetonitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Flash Point 101°C (213°F)
Boiling Point 110° to 114°C (14mmHg)
Quantity 100g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.