Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Thioisochroman-4-one, 98%

$84.04 - $287.85

Chemical Identifiers

Chemical Name or Material 2-Thioisochroman-4-one
CAS 4426-76-0
Molecular Formula C9H8OS
Formula Weight 164.23
MDL Number MFCD00051672
Beilstein 125059
Synonym 1H-2-Benzothiopyran-4(3H)-one; Isothiochroman-4-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0794803

  SDS
alfa aesar™
L0794803
1g Each for $84.04

AAL0794806

  SDS
alfa aesar™
L0794806
5g Each for $287.85
Description & Specifications

Specifications

Formula Weight 164.23
Chemical Name or Material 2-Thioisochroman-4-one
Assay 98%
Molecular Formula C9H8OS
MDL Number MFCD00051672
Beilstein 125059
Synonym 1H-2-Benzothiopyran-4(3H)-one; Isothiochroman-4-one
Melting Point 56° to 59°C
CAS 4426-76-0
Quantity 1g