Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Thiohydantoin, 99%

$34.18 - $400.22

Chemical Identifiers

Chemical Name or Material 2-Thiohydantoin
CAS 503-87-7
Molecular Formula C3H4N2OS
Formula Weight 116.14
MDL Number MFCD00005277
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2493406

 
alfa aesar™
B2493406
5g Each for $34.18

AAB2493414

 
alfa aesar™
B2493414
25g Each for $115.59

AAB2493422

 
alfa aesar™
B2493422
100g Each for $400.22
Description & Specifications

Specifications

Formula Weight 116.14
Chemical Name or Material 2-Thiohydantoin
TSCA TSCA
Assay 99%
Molecular Formula C3H4N2OS
MDL Number MFCD00005277
CAS 503-87-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number MU4200000
Melting Point 229° to 231°C (dec.)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.