missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Thiohydantoin, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 138761000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Thiohydantoin | |
Authentic | |
C3H4N2OS | |
100g | |
2-thiohydantoin, thiohydantoin, 4-imidazolidinone, 2-thioxo, 2-thioxoimidazolidin-4-one, 2-thioguidanthion, hydantoin, 2-thio, usaf be-25, 2-thioxo-4-imidazolidinone, 2-thioxo-1,3-diazolidin-4-one, glycine thiohydantoin | |
UGWULZWUXSCWPX-UHFFFAOYSA-N | |
2-sulfanylideneimidazolidin-4-one | |
1274030 | |
98% |
503-87-7 | |
97.5% min. (HPLC) | |
MFCD00005277 | |
24,26 | |
Solubility in water: slightly. | |
O=C1CNC(=S)N1 | |
116.14 | |
116.14 |
Chemical Identifiers
503-87-7 | |
116.14 | |
UGWULZWUXSCWPX-UHFFFAOYSA-N | |
1274030 | |
O=C1CNC(=S)N1 |
C3H4N2OS | |
MFCD00005277 | |
2-thiohydantoin, thiohydantoin, 4-imidazolidinone, 2-thioxo, 2-thioxoimidazolidin-4-one, 2-thioguidanthion, hydantoin, 2-thio, usaf be-25, 2-thioxo-4-imidazolidinone, 2-thioxo-1,3-diazolidin-4-one, glycine thiohydantoin | |
2-sulfanylideneimidazolidin-4-one |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
GHS Signal Word: Warning
EINECSNumber : 207-977-4
RUO â Research Use Only