Please login to your online account to display your discounted pricing

2-Thien-2-ylpyrimidine-5-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Thien-2-ylpyrimidine-5-carboxylic acid
CAS 916766-97-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H6N2O2S
Formula Weight 206.22
MDL Number MFCD09817551
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC66501DA

  SDS
fisher scientific company
CC66501DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.22
Quantity 1g
Assay 97%
CAS Max % 100.0
CAS 916766-97-7
Molecular Formula C9H6N2O2S
Chemical Name or Material 2-Thien-2-ylpyrimidine-5-carboxylic acid
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
MDL Number MFCD09817551
CAS Min % 96.5
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.