Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Thiazolylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material 2-Thiazolylzinc bromide
CAS 173382-28-0
Formula Weight 229.4
MDL Number MFCD00671987
UN Number UN3399
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH58497AD

  SDS
alfa aesar™
H58497AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.4
Chemical Name or Material 2-Thiazolylzinc bromide
Concentration 0.5M in THF
CAS 173382-28-0
MDL Number MFCD00671987
UN Number UN3399
Storage Note 1 2° to 8°C
Packaging ChemSeal™ bottles
Physical Form Liquid
Sensitivity Air Sensitive
Quantity 50mL
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Color Yellow to brown to black
Health Hazard 2 GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Density 0.992g/mL

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

2° to 8°C