Please login to your online account to display your discounted pricing

2-Thiazolecarboxaldehyde, 98%, ACROS Organics™


Quantity 250mg
Packaging Glass Ampule with Screw-cap

Manufacturer: thermo fisher scientific  AC377682500

 View more versions of this product

Catalog No. AC377682500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 2-Thiazolecarboxaldehyde
Density 1.2880g/mL
Color Clear pale yellow to brown
Quantity 250mg
CAS 10200-59-6
Health Hazard 1 Warning
Infrared Spectrum Authentic
Packaging Glass Ampule with Screw-cap
Physical Form Liquid
Assay ≥97.5% (GC)
Assay Percent Range 98%
Refractive Index 1.5735 to 1.5755 (20°C, 589nm)
Molecular Formula C4H3NOS
Formula Weight 113.14

Specifications

Chemical Name or Material 2-Thiazolecarboxaldehyde
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C4H3NOS
Formula Weight 113.14
Accessories

Warning

SDS & Safety Documents