missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Tetrazol-5-yl)phenylboronic acid, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 445550050

 View more versions of this product

Catalog No. AC445550050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-(Tetrazol-5-yl)phenylboronic acid
94% min. (HPLC)
5g
GVRXWYFECKHTSJ-UHFFFAOYSA-N
11171482
95%
155884-01-8
C7H7BN4O2
2-5-tetrazolyl phenylboronic acid, 2-1h-tetrazol-5-yl phenyl boronic acid, 2-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl-phenylboronic acid, 2-1h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenyl boronic acid, boronic acid, 2-1h-tetrazol-5-yl phenyl, boronic acid,b-2-2h-tetrazol-5-yl phenyl
189.97
189.97

Chemical Identifiers

155884-01-8
189.97
2-5-tetrazolyl phenylboronic acid, 2-1h-tetrazol-5-yl phenyl boronic acid, 2-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl-phenylboronic acid, 2-1h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenyl boronic acid, boronic acid, 2-1h-tetrazol-5-yl phenyl, boronic acid,b-2-2h-tetrazol-5-yl phenyl
C7H7BN4O2
GVRXWYFECKHTSJ-UHFFFAOYSA-N
11171482
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only