Please login to your online account to display your discounted pricing

2-(Tetrahydropyran-4-yloxy)benzoyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(Tetrahydropyran-4-yloxy)benzoyl chloride
CAS 906352-68-9
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C12H13ClO3
Formula Weight 240.68
MDL Number MFCD09025870
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC52202CB

 
fisher scientific company
CC52202CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.68
Quantity 250mg
CAS Max % 100.0
Packaging Amber Glass Bottle
CAS Min % 94.5
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 2-(Tetrahydropyran-4-yloxy)benzoyl chloride
Assay Percent Range 95%
Molecular Formula C12H13ClO3
MDL Number MFCD09025870
CAS 906352-68-9
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.