missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Tetrahydropyran-4-ylethanol, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company CC67409CB

 View more versions of this product

Catalog No. CC67409CB



Description

Description

Specifications

Specifications

2-Tetrahydropyran-4-ylethanol
250mg
96.5
97%
MFCD00129068
XZXZZACRGBBWTQ-UHFFFAOYSA-N
2-(oxan-4-yl)ethanol
17750944
97%
Amber Glass Bottle
4677-18-3
100.0
C7H14O2
2-oxan-4-yl ethan-1-ol, 2-oxan-4-yl ethanol, 2-tetrahydro-2h-pyran-4-yl ethan-1-ol, 2-tetrahydro-2h-pyran-4-yl ethanol, 2-tetrahydro-2h-pyran-4-ylethanol, 2-tetrahydro-pyran-4-yl-ethanol, 2-tetrahydropyran-4-ylethanol, 2h-pyran-4-ethanol, tetrahydro, 4-2-hydroxyethyl oxane, 4-hydroxyethyl tetrahydropyran
C1COCCC1CCO
130.187
130.19

Chemical Identifiers

4677-18-3
130.187
XZXZZACRGBBWTQ-UHFFFAOYSA-N
17750944
C1COCCC1CCO
C7H14O2
MFCD00129068
2-oxan-4-yl ethan-1-ol, 2-oxan-4-yl ethanol, 2-tetrahydro-2h-pyran-4-yl ethan-1-ol, 2-tetrahydro-2h-pyran-4-yl ethanol, 2-tetrahydro-2h-pyran-4-ylethanol, 2-tetrahydro-pyran-4-yl-ethanol, 2-tetrahydropyran-4-ylethanol, 2h-pyran-4-ethanol, tetrahydro, 4-2-hydroxyethyl oxane, 4-hydroxyethyl tetrahydropyran
2-(oxan-4-yl)ethanol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents