missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Tetrahydrofurfuryloxy)tetrahydropyran 98%, ACROS Organics™

Chemical Identifiers

CAS 710-14-5
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.251
MDL Number MFCD00006603
InChI Key XXNUDHUOMVIIFN-UHFFFAOYSA-N
Synonym 2 3-tetrahydrofurfuryloxy tetrahydro-pyran, 2 3-tetrahydrofurfuryloxy tetrahydropyran, 2-oxolan-2-ylmethoxy oxane, 2-tetrahydrofurfuroxy tetrahydropyran, 2-tetrahydrofurfuryloxy tetrahydropyran, 2h-pyran, tetrahydro-2-tetrahydro-2-furanyl methoxy, 2h-pyran, tetrahydro-2-tetrahydrofurfuryl oxy, acmc-209soh, tetrahydro tetrahydrofuryl methoxy-2h-pyran, tetrahydrofurfuroxy tetrahydropyran
PubChem CID 97985
IUPAC Name 2-(oxolan-2-ylmethoxy)oxane
SMILES C1CCOC(C1)OCC2CCCO2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC138190250
View Documents
Thermo Fisher Scientific
138190250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

710-14-5
186.251
XXNUDHUOMVIIFN-UHFFFAOYSA-N
97985
C1CCOC(C1)OCC2CCCO2
C10H18O3
MFCD00006603
2 3-tetrahydrofurfuryloxy tetrahydro-pyran, 2 3-tetrahydrofurfuryloxy tetrahydropyran, 2-oxolan-2-ylmethoxy oxane, 2-tetrahydrofurfuroxy tetrahydropyran, 2-tetrahydrofurfuryloxy tetrahydropyran, 2h-pyran, tetrahydro-2-tetrahydro-2-furanyl methoxy, 2h-pyran, tetrahydro-2-tetrahydrofurfuryl oxy, acmc-209soh, tetrahydro tetrahydrofuryl methoxy-2h-pyran, tetrahydrofurfuroxy tetrahydropyran
2-(oxolan-2-ylmethoxy)oxane

Specifications

1.03g/mL
Glass bottle
Colorless
710-14-5
100.0
C10H18O3
2 3-tetrahydrofurfuryloxy tetrahydro-pyran, 2 3-tetrahydrofurfuryloxy tetrahydropyran, 2-oxolan-2-ylmethoxy oxane, 2-tetrahydrofurfuroxy tetrahydropyran, 2-tetrahydrofurfuryloxy tetrahydropyran, 2h-pyran, tetrahydro-2-tetrahydro-2-furanyl methoxy, 2h-pyran, tetrahydro-2-tetrahydrofurfuryl oxy, acmc-209soh, tetrahydro tetrahydrofuryl methoxy-2h-pyran, tetrahydrofurfuroxy tetrahydropyran
C1CCOC(C1)OCC2CCCO2
186.251
186.25
≥97.5%
Authentic
1.4600 to 1.4620 (20°C, 589nm)
25g
97.5
98%
MFCD00006603
XXNUDHUOMVIIFN-UHFFFAOYSA-N
2-(oxolan-2-ylmethoxy)oxane
97985
Liquid
2-(Tetrahydrofurfuryloxy)tetrahydropyran, 98%
Safety and Handling

Safety and Handling

EINECSNumber : 211-917-2

TSCA : TSCA

SDS