Please login to your online account to display your discounted pricing

2-(Tetrahydrofurfuryloxy)tetrahydropyran 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  138191000

 View more versions of this product

Catalog No. AC138191000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 186.25
Quantity 100g
Color Clear colorless
EINECS Number 211-917-2
TSCA TSCA
Density 1.03g/mL
CAS Max % 100.0
Chemical Name or Material 2-(Tetrahydrofurfuryloxy)tetrahydropyran
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10H18O3
CAS 710-14-5
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4600 to 1.4620 (20°C, 589nm)
MDL Number MFCD00006603

Specifications

Chemical Name or Material 2-(Tetrahydrofurfuryloxy)tetrahydropyran
Name Note 98%
CAS 710-14-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H18O3
Formula Weight 186.25
MDL Number MFCD00006603