Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(tert-Butylthio)benzaldehyde, 96%

$32.05 - $32.05

Chemical Identifiers

Chemical Name or Material 2-(tert-Butylthio)benzaldehyde
CAS 65924-65-4
Molecular Formula C11H14OS
Formula Weight 194.30g/mol
MDL Number MFCD00040294
Synonym 2-(tert-Butylmercapto)benzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1552403

 
alfa aesar™
A15524-03
1g Each for $32.05
Description & Specifications

Specifications

Boiling Point 95° to 97°C (0.1mmHg)
Quantity 1g
Chemical Name or Material 2-(tert-Butylthio)benzaldehyde
CAS 65924-65-4
Assay 96%
Molecular Formula C11H14OS
Formula Weight 194.30g/mol
MDL Number MFCD00040294
Synonym 2-(tert-Butylmercapto)benzaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.