Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(tert-Butylsulfonyl)thioacetamide, 97%

$159.72 - $721.74

Chemical Identifiers

Chemical Name or Material 2-(tert-Butylsulfonyl)thioacetamide
CAS 175277-31-3
Molecular Formula C6H13NO2S2
Formula Weight 195.29
MDL Number MFCD00084920
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2670603

  SDS
alfa aesar™
H2670603
1g Each for $159.72

AAH2670606

  SDS
alfa aesar™
H2670606
5g Each for $721.74
Description & Specifications

Specifications

Formula Weight 195.29
MDL Number MFCD00084920
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 175277-31-3
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C6H13NO2S2
Quantity 1g
Melting Point 184° to 186°C
Chemical Name or Material 2-(tert-Butylsulfonyl)thioacetamide
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.