Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-tert-Butylpropane-1,3-diol, 98%

$64.18 - $219.99

Chemical Identifiers

Chemical Name or Material 2-tert-Butylpropane-1,3-diol
CAS 2819-05-8
Molecular Formula C7H16O2
Formula Weight 132.2
MDL Number MFCD01075745
Beilstein 1900687
Synonym 3,3-Dimethyl-2-(hydroxymethyl)-1-butanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1692703

  SDS
alfa aesar™
L1692703
1g Each for $64.18

AAL1692706

  SDS
alfa aesar™
L1692706
5g Each for $219.99
Description & Specifications

Specifications

Formula Weight 132.2
Chemical Name or Material 2-tert-Butylpropane-1,3-diol
CAS 2819-05-8
Assay 98%
Molecular Formula C7H16O2
MDL Number MFCD01075745
Beilstein 1900687
Synonym 3,3-Dimethyl-2-(hydroxymethyl)-1-butanol
Boiling Point 100° to 102°C (2mmHg)
Melting Point 54° to 56°C
Quantity 1g
RTECS Number TY3540020