Learn More
2-tert-Butylphenol, 99%, Thermo Scientific Chemicals
$50.21 - $180.58
Chemical Identifiers
CAS | 88-18-6 |
---|---|
Molecular Formula | C10H14O |
Molecular Weight (g/mol) | 150.221 |
MDL Number | MFCD00002223 |
InChI Key | WJQOZHYUIDYNHM-UHFFFAOYSA-N |
Synonym | 2-tert-butyl phenol, 2-t-butylphenol, phenol, 2-1,1-dimethylethyl, phenol, o-tert-butyl, o-t-butylphenol, o-tert-butylphenol, tert-butylphenol, 2-1,1-dimethylethyl phenol, 2-tert-butyl-1-hydroxybenzene, t-butylphenol |
PubChem CID | 6923 |
ChEBI | CHEBI:34305 |
IUPAC Name | 2-tert-butylphenol |
SMILES | CC(C)(C)C1=CC=CC=C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0729422
|
Thermo Scientific Chemicals
L0729422 |
100 g |
Each for $50.21
|
|
|||||
AAL072940P
|
Thermo Scientific Chemicals
L072940P |
2000 g |
Each for $180.58
|
|
|||||
Description
2-tert-Butylphenol is used pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-tert-Butylphenol is used pharmaceutical intermediates.
Solubility
Soluble in water (0.23 g/100 mL (20°C)).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
88-18-6 | |
150.221 | |
WJQOZHYUIDYNHM-UHFFFAOYSA-N | |
6923 | |
2-tert-butylphenol |
C10H14O | |
MFCD00002223 | |
2-tert-butyl phenol, 2-t-butylphenol, phenol, 2-1,1-dimethylethyl, phenol, o-tert-butyl, o-t-butylphenol, o-tert-butylphenol, tert-butylphenol, 2-1,1-dimethylethyl phenol, 2-tert-butyl-1-hydroxybenzene, t-butylphenol | |
CHEBI:34305 | |
CC(C)(C)C1=CC=CC=C1O |
Specifications
88-18-6 | |
0.98 | |
102°C (215°F) | |
1.523 | |
100 g | |
1907120 | |
Soluble in water (0.23g/100 mL (20°C)). | |
CC(C)(C)C1=CC=CC=C1O | |
150.221 | |
CHEBI:34305 | |
99% |
-7°C | |
223°C to 224°C | |
C10H14O | |
MFCD00002223 | |
UN2922 | |
2-tert-butyl phenol, 2-t-butylphenol, phenol, 2-1,1-dimethylethyl, phenol, o-tert-butyl, o-t-butylphenol, o-tert-butylphenol, tert-butylphenol, 2-1,1-dimethylethyl phenol, 2-tert-butyl-1-hydroxybenzene, t-butylphenol | |
WJQOZHYUIDYNHM-UHFFFAOYSA-N | |
2-tert-butylphenol | |
6923 | |
150.22 | |
2-tert-Butylphenol |
Safety and Handling
GHS H Statement
H311-H330-H314-H318-H302
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H331-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 201-807-2
RTECSNumber : SJ8921000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only