Learn More
2-tert-Butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 439160010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-tert-Butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine | |
97.5 | |
0.9480g/mL | |
53°C | |
97.5% min. (GC) | |
C13H31N4P | |
0.948 | |
VSCBATMPTLKTOV-UHFFFAOYSA-N | |
2-tert-butylimino-N,N-diethyl-1,3-dimethyl-1,3,2$l^{5}-diazaphosphinan-2-amine | |
3513851 | |
98% |
98015-45-3 | |
100.0 | |
74°C (0.1mmHg) | |
Authentic | |
Glass bottle | |
1mL | |
bemp, bemp phosphazene, 2-tert-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, unii-df15j146qi, 2-tert-butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine, 2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2??-diazaphosphinan-2-amine, 2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2, 2-t-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, 2-tert-butylimino-2-diethylamino-1,3-dimethyl-1,3,2-diazaphosphorinane | |
CCN(CC)P1(=NC(C)(C)C)N(CCCN1C)C | |
274.39 | |
274.39 |
Chemical Identifiers
98015-45-3 | |
274.39 | |
bemp, bemp phosphazene, 2-tert-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, unii-df15j146qi, 2-tert-butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine, 2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2??-diazaphosphinan-2-amine, 2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2, 2-t-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, 2-tert-butylimino-2-diethylamino-1,3-dimethyl-1,3,2-diazaphosphorinane | |
2-tert-butylimino-N,N-diethyl-1,3-dimethyl-1,3,2$l^{5}-diazaphosphinan-2-amine |
C13H31N4P | |
VSCBATMPTLKTOV-UHFFFAOYSA-N | |
3513851 | |
CCN(CC)P1(=NC(C)(C)C)N(CCCN1C)C |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im
GHS Signal Word: Danger
RUO â Research Use Only