missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(tert-Butyldimethylsilyl)thiazole, TCI America™

$99.31 - $340.70

Chemical Identifiers

CAS 137382-38-8
Molecular Formula C9H17NSSi
Molecular Weight (g/mol) 199.387
MDL Number MFCD10699169
InChI Key DKQFKRXPJSBPOF-UHFFFAOYSA-N
PubChem CID 10465374
IUPAC Name tert-butyl-dimethyl-(1,3-thiazol-2-yl)silane
SMILES CC(C)(C)[Si](C)(C)C1=NC=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B46541G View Documents TCI America
B46541G
1g Each for $99.31
 
B46545G View Documents TCI America
B46545G
5g Each for $340.70
 
Specifications

Chemical Identifiers

137382-38-8
199.387
DKQFKRXPJSBPOF-UHFFFAOYSA-N
tert-butyl-dimethyl-(1,3-thiazol-2-yl)silane
C9H17NSSi
MFCD10699169
10465374
CC(C)(C)[Si](C)(C)C1=NC=CS1

Specifications

Yellow
137382-38-8
MFCD10699169
CC(C)(C)[Si](C)(C)C1=NC=CS1
199.387
199.39
≥97.0% (GC)
1g
C9H17NSSi
DKQFKRXPJSBPOF-UHFFFAOYSA-N
tert-butyl-dimethyl-(1,3-thiazol-2-yl)silane
10465374
Clear Liquid at 20°C
2-(tert-Butyldimethylsilyl)thiazole
Safety and Handling

Safety and Handling

TSCA : No