Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-tert-Butylanthraquinone, 95%

$97.58 - $97.58

Chemical Identifiers

Chemical Name or Material 2-tert-Butylanthraquinone
CAS 84-47-9
Molecular Formula C18H16O2
Formula Weight 264.32g/mol
MDL Number MFCD00001232
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2363322

 
alfa aesar™
B23633-22
100g Each for $97.58
Description & Specifications

Specifications

Melting Point 98° to 100°C
Quantity 100g
Chemical Name or Material 2-tert-Butylanthraquinone
CAS 84-47-9
Assay 95%
Molecular Formula C18H16O2
Formula Weight 264.32g/mol
MDL Number MFCD00001232
RTECS Number CB0487500
TSCA TSCA