missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(tert-Butylamino)ethanol, TCI America™

$30.56 - $30.56

Chemical Identifiers

CAS 4620-70-6
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00002837
InChI Key IUXYVKZUDNLISR-UHFFFAOYSA-N
Synonym 2-tert-butylamino ethanol, n-tert-butylethanolamine, tert-butylaminoethanol, t-butylethanolamine, ethanol, 2-tert-butylamino, ethanol, 2-1,1-dimethylethyl amino, n-t-butylethanolamine, 2-t-butylamino ethanol, 2-1,1-dimethylethyl amino ethanol, .beta.-tert-butylaminoethanol
PubChem CID 78361
IUPAC Name 2-(tert-butylamino)ethanol
SMILES CC(C)(C)NCCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B110125G View Documents TCI America
B110125G
25g Each for $30.56
 
Specifications

Chemical Identifiers

4620-70-6
117.192
IUXYVKZUDNLISR-UHFFFAOYSA-N
78361
CC(C)(C)NCCO
C6H15NO
MFCD00002837
2-tert-butylamino ethanol, n-tert-butylethanolamine, tert-butylaminoethanol, t-butylethanolamine, ethanol, 2-tert-butylamino, ethanol, 2-1,1-dimethylethyl amino, n-t-butylethanolamine, 2-t-butylamino ethanol, 2-1,1-dimethylethyl amino ethanol, .beta.-tert-butylaminoethanol
2-(tert-butylamino)ethanol

Specifications

177°C
42°C
4620-70-6
MFCD00002837
2-tert-butylamino ethanol, n-tert-butylethanolamine, tert-butylaminoethanol, t-butylethanolamine, ethanol, 2-tert-butylamino, ethanol, 2-1,1-dimethylethyl amino, n-t-butylethanolamine, 2-t-butylamino ethanol, 2-1,1-dimethylethyl amino ethanol, .beta.-tert-butylaminoethanol
CC(C)(C)NCCO
117.192
117.19
≥98.0% (GC,T)
White-Yellow
25g
C6H15NO
2923
IUXYVKZUDNLISR-UHFFFAOYSA-N
2-(tert-butylamino)ethanol
78361
Crystal-Lump at 20°C
2-(tert-Butylamino)ethanol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0295

RTECSNumber : KK7177500

TSCA : Yes