missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-1,3,5-triazine, TCI America™

$111.53 - $381.95

Chemical Identifiers

CAS 28159-98-0
Molecular Formula C11H19N5S
Molecular Weight (g/mol) 253.368
MDL Number MFCD01863779
InChI Key HDHLIWCXDDZUFH-UHFFFAOYSA-N
Synonym Cybutryne
PubChem CID 91590
ChEBI CHEBI:5962
IUPAC Name 2-N-tert-butyl-4-N-cyclopropyl-6-methylsulfanyl-1,3,5-triazine-2,4-diamine
SMILES CC(C)(C)NC1=NC(=NC(=N1)NC2CC2)SC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I08425G View Documents TCI America
I08425G
5g Each for $111.53
 
I084225G View Documents TCI America
I084225G
25g Each for $381.95
 
Specifications

Chemical Identifiers

28159-98-0
253.368
HDHLIWCXDDZUFH-UHFFFAOYSA-N
91590
2-N-tert-butyl-4-N-cyclopropyl-6-methylsulfanyl-1,3,5-triazine-2,4-diamine
C11H19N5S
MFCD01863779
Cybutryne
CHEBI:5962
CC(C)(C)NC1=NC(=NC(=N1)NC2CC2)SC

Specifications

White
5g
C11H19N5S
3077
HDHLIWCXDDZUFH-UHFFFAOYSA-N
2-N-tert-butyl-4-N-cyclopropyl-6-methylsulfanyl-1,3,5-triazine-2,4-diamine
91590
253.37
≥98.0% (T)
128°C
28159-98-0
MFCD01863779
Cybutryne
CC(C)(C)NC1=NC(=NC(=N1)NC2CC2)SC
253.368
CHEBI:5962
Crystal-Powder at 20°C
2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-1,3,5-triazine
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6110

RTECSNumber : XY5850675

TSCA : No