missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-tert-Butyl-4-methylphenol 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 107965000

 View more versions of this product

Catalog No. AC107965000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-tert-Butyl-4-methylphenol
2409-55-4
100.0
Crystalline Mass
99%
(CH3)3CC6H3(CH3)OH
164.25
2-tert-Butyl-p-cresol
White to yellow-beige
99%
98.5
Authentic
≥98.5%
C11H16O
Glass bottle
MFCD00002381
50° to 53°C
500g

Chemical Identifiers

2409-55-4
100.0
C11H16O
164.25
2-tert-Butyl-p-cresol
≥98.5%
98.5
99%
(CH3)3CC6H3(CH3)OH
MFCD00002381
2-tert-Butyl-4-methylphenol, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 219-314-6

RTECSNumber : GO7000000

TSCA : TSCA