Please login to your online account to display your discounted pricing

2-(tert-Butoxycarbonylamino)phenylboronic acid pinacol ester, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC444650010

 View more versions of this product

Catalog No. AC444650010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 319.21
Quantity 1g
Chemical Name or Material 2-(tert-Butoxycarbonylamino)phenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C17H26BNO4
Synonym 2-(N-BOC-Amino)phenylboronic acid pinacol ester
Health Hazard 1 Warning

Specifications

Chemical Name or Material 2-(tert-Butoxycarbonylamino)phenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C17H26BNO4
Formula Weight 319.21
Synonym 2-(N-BOC-Amino)phenylboronic acid pinacol ester
Accessories

Warning