Please login to your online account to display your discounted pricing

2-(tert-Butoxycarbonylamino)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(tert-Butoxycarbonylamino)phenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C17H26BNO4
Formula Weight 319.21
Synonym 2-(N-BOC-Amino)phenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444650050

 
thermo fisher scientific
AC444650050
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444650010

 
thermo fisher scientific
AC444650010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 319.21
Quantity 1g
Chemical Name or Material 2-(tert-Butoxycarbonylamino)phenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C17H26BNO4
Synonym 2-(N-BOC-Amino)phenylboronic acid pinacol ester
Health Hazard 1 Warning
Safety and Handling

Warning