missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(tert-Butoxycarbonylamino)phenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 146140-95-6
Molecular Formula C11H16BNO3
Molecular Weight (g/mol) 221.06
MDL Number MFCD01114645
InChI Key MXRAJVMTCAUABO-UHFFFAOYSA-N
Synonym 2-2,2,2-trimethylacetamido benzeneboronic acid, 2-2,2-dimethyl-propionylamino phenylboronic acid, 2-2,2-dimethylpropanamido phenylboronic acid, 2-2,2-dimethylpropanoylamino phenyl boronic acid, 2-pivalamido phenylboronic acid, 2-pivalamidophenyl boronic acid, 2-pivaloylamino phenylboronic acid, 2-pivaloylaminobenzene boronic acid, 2-tert-butylcarbonylamino phenylboronic acid, boronic acid,b-2-2,2-dimethyl-1-oxopropyl amino phenyl
PubChem CID 4193502
IUPAC Name [2-(2,2-dimethylpropanoylamino)phenyl]boronic acid
SMILES CC(C)(C)C(=O)NC1=CC=CC=C1B(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445340010
SDS View Documents
Thermo Fisher Scientific
AC445340010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445340050
SDS View Documents
Thermo Fisher Scientific
AC445340050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

146140-95-6
221.06
MXRAJVMTCAUABO-UHFFFAOYSA-N
4193502
CC(C)(C)C(=O)NC1=CC=CC=C1B(O)O
C11H16BNO3
MFCD01114645
2-2,2,2-trimethylacetamido benzeneboronic acid, 2-2,2-dimethyl-propionylamino phenylboronic acid, 2-2,2-dimethylpropanamido phenylboronic acid, 2-2,2-dimethylpropanoylamino phenyl boronic acid, 2-pivalamido phenylboronic acid, 2-pivalamidophenyl boronic acid, 2-pivaloylamino phenylboronic acid, 2-pivaloylaminobenzene boronic acid, 2-tert-butylcarbonylamino phenylboronic acid, boronic acid,b-2-2,2-dimethyl-1-oxopropyl amino phenyl
[2-(2,2-dimethylpropanoylamino)phenyl]boronic acid

Specifications

Glass Bottle
1g
96% min. (HPLC)
MFCD01114645
MXRAJVMTCAUABO-UHFFFAOYSA-N
[2-(2,2-dimethylpropanoylamino)phenyl]boronic acid
4193502
97%
296.5°C to 297.5°C
146140-95-6
C11H16BNO3
2-2,2,2-trimethylacetamido benzeneboronic acid, 2-2,2-dimethyl-propionylamino phenylboronic acid, 2-2,2-dimethylpropanamido phenylboronic acid, 2-2,2-dimethylpropanoylamino phenyl boronic acid, 2-pivalamido phenylboronic acid, 2-pivalamidophenyl boronic acid, 2-pivaloylamino phenylboronic acid, 2-pivaloylaminobenzene boronic acid, 2-tert-butylcarbonylamino phenylboronic acid, boronic acid,b-2-2,2-dimethyl-1-oxopropyl amino phenyl
CC(C)(C)C(=O)NC1=CC=CC=C1B(O)O
221.06
221.06
2-(tert-Butoxycarbonylamino)phenylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS