Learn More
2-(tert-Butoxycarbonylamino)phenylboronic acid, 97%, Thermo Scientific™
Supplier: Thermo Fisher Scientific 445340050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-(tert-Butoxycarbonylamino)phenylboronic acid | |
296.5°C to 297.5°C | |
C11H16BNO3 | |
5g | |
MXRAJVMTCAUABO-UHFFFAOYSA-N | |
[2-(2,2-dimethylpropanoylamino)phenyl]boronic acid | |
4193502 | |
97% |
146140-95-6 | |
96% min. (HPLC) | |
MFCD01114645 | |
2-2,2,2-trimethylacetamido benzeneboronic acid, 2-2,2-dimethyl-propionylamino phenylboronic acid, 2-2,2-dimethylpropanamido phenylboronic acid, 2-2,2-dimethylpropanoylamino phenyl boronic acid, 2-pivalamido phenylboronic acid, 2-pivalamidophenyl boronic acid, 2-pivaloylamino phenylboronic acid, 2-pivaloylaminobenzene boronic acid, 2-tert-butylcarbonylamino phenylboronic acid, boronic acid,b-2-2,2-dimethyl-1-oxopropyl amino phenyl | |
CC(C)(C)C(=O)NC1=CC=CC=C1B(O)O | |
221.06 | |
221.06 |
Chemical Identifiers
146140-95-6 | |
221.06 | |
MXRAJVMTCAUABO-UHFFFAOYSA-N | |
4193502 | |
CC(C)(C)C(=O)NC1=CC=CC=C1B(O)O |
C11H16BNO3 | |
MFCD01114645 | |
2-2,2,2-trimethylacetamido benzeneboronic acid, 2-2,2-dimethyl-propionylamino phenylboronic acid, 2-2,2-dimethylpropanamido phenylboronic acid, 2-2,2-dimethylpropanoylamino phenyl boronic acid, 2-pivalamido phenylboronic acid, 2-pivalamidophenyl boronic acid, 2-pivaloylamino phenylboronic acid, 2-pivaloylaminobenzene boronic acid, 2-tert-butylcarbonylamino phenylboronic acid, boronic acid,b-2-2,2-dimethyl-1-oxopropyl amino phenyl | |
[2-(2,2-dimethylpropanoylamino)phenyl]boronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only