missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(tert-Butoxycarbonylamino)ethyl Bromide 98.0+%, TCI America™
$109.49 - $985.44
Chemical Identifiers
CAS | 39684-80-5 |
---|---|
Molecular Formula | C7H14BrNO2 |
Molecular Weight (g/mol) | 224.10 |
MDL Number | MFCD02683428 |
InChI Key | TZRQZPMQUXEZMC-UHFFFAOYSA-N |
Synonym | tert-butyl n-2-bromoethyl carbamate, 2-boc-amino ethyl bromide, n-boc-bromoethylamine, tert-butyl 2-bromoethyl carbamate, n-boc-2-bromoethylamine, tert-butyl 2-bromoethylcarbamate, 2-bromo-ethyl-carbamic acid tert-butyl ester, 2-tert-butoxycarbonylamino ethyl bromide, 2-boc-amino-ethyl bromide |
PubChem CID | 4103526 |
IUPAC Name | tert-butyl N-(2-bromoethyl)carbamate |
SMILES | CC(C)(C)OC(=O)NCCBr |
Chemical Identifiers
39684-80-5 | |
224.10 | |
TZRQZPMQUXEZMC-UHFFFAOYSA-N | |
4103526 | |
CC(C)(C)OC(=O)NCCBr |
C7H14BrNO2 | |
MFCD02683428 | |
tert-butyl n-2-bromoethyl carbamate, 2-boc-amino ethyl bromide, n-boc-bromoethylamine, tert-butyl 2-bromoethyl carbamate, n-boc-2-bromoethylamine, tert-butyl 2-bromoethylcarbamate, 2-bromo-ethyl-carbamic acid tert-butyl ester, 2-tert-butoxycarbonylamino ethyl bromide, 2-boc-amino-ethyl bromide | |
tert-butyl N-(2-bromoethyl)carbamate |
Specifications
33°C | |
White-Yellow | |
MFCD02683428 | |
TZRQZPMQUXEZMC-UHFFFAOYSA-N | |
tert-butyl N-(2-bromoethyl)carbamate | |
4103526 | |
≥98.0% (GC) | |
Crystalline Powder |
39684-80-5 | |
C7H14BrNO2 | |
tert-butyl n-2-bromoethyl carbamate, 2-boc-amino ethyl bromide, n-boc-bromoethylamine, tert-butyl 2-bromoethyl carbamate, n-boc-2-bromoethylamine, tert-butyl 2-bromoethylcarbamate, 2-bromo-ethyl-carbamic acid tert-butyl ester, 2-tert-butoxycarbonylamino ethyl bromide, 2-boc-amino-ethyl bromide | |
CC(C)(C)OC(=O)NCCBr | |
224.10 | |
224.10 | |
1 g | |
2-(tert-Butoxycarbonylamino)ethyl Bromide |
Safety and Handling
TSCA : No
Recommended Storage : Freezer