missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(tert-Butoxycarbonylamino)-4-methylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

CAS 1150561-75-3
Molecular Formula C18H28BNO4
Molecular Weight (g/mol) 333.24
Synonym 2-(N-BOC-Amino)-4-methylphenylboronic acid pinacol ester
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444630010 SDS View Documents Thermo Fisher Scientific
AC444630010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC444630050 SDS View Documents Thermo Fisher Scientific
AC444630050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1150561-75-3
C18H28BNO4
333.24
2-(N-BOC-Amino)-4-methylphenylboronic acid pinacol ester

Specifications

Glass Bottle
1g
97%
2-(N-BOC-Amino)-4-methylphenylboronic acid pinacol ester
333.24
2-(tert-Butoxycarbonylamino)-4-methylphenylboronic acid pinacol ester
128°C to 130°C
1150561-75-3
C18H28BNO4
333.24
97%
Safety and Handling

Safety and Handling

GHS Signal Word: Warning