missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-tert-Butoxy-6-chloropyridine, 97%

Manufacturer:  Alfa Aesar™ H5508514

 View more versions of this product

Catalog No. AAH5508514



Specifications

Specifications

2-tert-Butoxy-6-chloropyridine
Colorless
547740-43-2
MFCD08705263
PLDRTFJRLUKTTA-UHFFFAOYSA-N
2-chloro-6-[(2-methylpropan-2-yl)oxy]pyridine
11984350
Liquid
1.076g/mL
25g
C9H12ClNO
2-tert-butoxy-6-chloro-pyridine
CC(C)(C)OC1=NC(=CC=C1)Cl
185.651
185.65
97%

Chemical Identifiers

547740-43-2
185.651
PLDRTFJRLUKTTA-UHFFFAOYSA-N
11984350
CC(C)(C)OC1=NC(=CC=C1)Cl
C9H12ClNO
MFCD08705263
2-tert-butoxy-6-chloro-pyridine
2-chloro-6-[(2-methylpropan-2-yl)oxy]pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents