missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Sulfobenzoic acid hydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2338122

 View more versions of this product

Catalog No. AAB2338122



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Sulfobenzoic acid hydrate
∼68°C to 72°C
MFCD00007479
UN3261
Hygroscopic
ZMPRRFPMMJQXPP-UHFFFAOYSA-N
2-sulfobenzoic acid;hydrate
24820382
98%
123333-68-6
C7H6O5S
100 g
2106852
2-sulfobenzoic acid hydrate, benzoic acid, 2-sulfo-hydrate, acmc-20anmu, 2-sulphobenzoic acid hydrate, c7h6o5s.h2o, 2-sulfobenzoic acid, oxamethane, benzoic acid, 2-sulfo-, monohydrate
OC(=O)C1=CC=CC=C1S(O)(=O)=O
202.18
202.19 (Anhydrous)

Chemical Identifiers

123333-68-6
202.18
ZMPRRFPMMJQXPP-UHFFFAOYSA-N
24820382
OC(=O)C1=CC=CC=C1S(O)(=O)=O
C7H6O5S
MFCD00007479
2-sulfobenzoic acid hydrate, benzoic acid, 2-sulfo-hydrate, acmc-20anmu, 2-sulphobenzoic acid hydrate, c7h6o5s.h2o, 2-sulfobenzoic acid, oxamethane, benzoic acid, 2-sulfo-, monohydrate
2-sulfobenzoic acid;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 211-176-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only