Learn More
2-Sulfobenzoic acid cyclic anhydride, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 184750250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Sulfobenzoic acid cyclic anhydride | |
81-08-3 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00005879 | |
2-sulfobenzoic anhydride, 2-sulfobenzoic acid cyclic anhydride, sulfobenzoic anhydride, o-sulfobenzoic acid anhydride, 3h-2,1-benzoxathiol-3-one, 1,1-dioxide, 3h-benzo c 1,2 oxathiol-3-one 1,1-dioxide, unii-0swh68iq5w, o-sulfobenzoic acid, cyclic anhydride, 2,1-benzoxathiol-3-one-1,1-dioxide, 0swh68iq5w | |
C1=CC=C2C(=C1)C(=O)OS2(=O)=O | |
184.17 | |
184.17 | |
Technical |
tech., 90% | |
94.0 | |
184°C to 186°C (18.0mmHg) | |
95% | |
C7H4O4S | |
25g | |
NCYNKWQXFADUOZ-UHFFFAOYSA-N | |
1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one | |
65729 | |
95% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
EINECSNumber : 201-322-6
TSCA : TSCA
RUO â Research Use Only