Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-sec-Butylthiazole, 98+%

Chemical Identifiers

Chemical Name or Material 2-sec-Butylthiazole
CAS 18277-27-5
Molecular Formula C7H11NS
Formula Weight 141.24
MDL Number MFCD00051951
UN Number UN1993
Synonym 2-(1-Methylpropyl)thiazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1348314

 
alfa aesar™
A1348314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1348322

 
alfa aesar™
A1348322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.24
Chemical Name or Material 2-sec-Butylthiazole
Assay ≥98%
Molecular Formula C7H11NS
MDL Number MFCD00051951
CAS 18277-27-5
UN Number UN1993
Synonym 2-(1-Methylpropyl)thiazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 174° to 175°C
Quantity 25g
Flash Point 58°C (136°F)
Odor Characteristic
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.