Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-sec-Butyl-4,5-dimethyl-3-thiazoline, 98%

$65.73 - $344.80

Chemical Identifiers

Chemical Name or Material 2-sec-Butyl-4,5-dimethyl-3-thiazoline
CAS 65894-82-8
Molecular Formula C9H17NS
Formula Weight 171.3
MDL Number MFCD01680858
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2687003

  SDS
alfa aesar™
H2687003
1g Each for $65.73

AAH2687009

  SDS
alfa aesar™
H2687009
10g Each for $344.80
Description & Specifications

Specifications

Formula Weight 171.3
Chemical Name or Material 2-sec-Butyl-4,5-dimethyl-3-thiazoline
Assay 98%
Molecular Formula C9H17NS
MDL Number MFCD01680858
CAS 65894-82-8
Boiling Point 71.00°C (4.00mmHg)
Quantity 1g
Flash Point >70°C (158°F)