missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Quinoxalinecarboxylic acid, 97%, Maybridge

Chemical Identifiers

CAS 879-65-2
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00012334
InChI Key UPUZGXILYFKSGE-UHFFFAOYSA-N
Synonym 2-quinoxalinecarboxylic acid, 2-quinoxalinecarboxylicacid, unii-m5oe8sn42m, 2-carboxyquinoxaline, m5oe8sn42m, 2-qca, pubchem9776, carbadox metabolite, acmc-1bl7g, quinoxaline carboxylic acid
PubChem CID 96695
IUPAC Name quinoxaline-2-carboxylic acid
SMILES C1=CC=C2C(=C1)N=CC(=N2)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC03501DE View Documents Fisher Scientific Company
CC03501DE
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

879-65-2
174.159
UPUZGXILYFKSGE-UHFFFAOYSA-N
96695
C1=CC=C2C(=C1)N=CC(=N2)C(=O)O
C9H6N2O2
MFCD00012334
2-quinoxalinecarboxylic acid, 2-quinoxalinecarboxylicacid, unii-m5oe8sn42m, 2-carboxyquinoxaline, m5oe8sn42m, 2-qca, pubchem9776, carbadox metabolite, acmc-1bl7g, quinoxaline carboxylic acid
quinoxaline-2-carboxylic acid

Specifications

5g
96.5
97%
MFCD00012334
UPUZGXILYFKSGE-UHFFFAOYSA-N
quinoxaline-2-carboxylic acid
96695
97%
879-65-2
100.0
C9H6N2O2
2-quinoxalinecarboxylic acid, 2-quinoxalinecarboxylicacid, unii-m5oe8sn42m, 2-carboxyquinoxaline, m5oe8sn42m, 2-qca, pubchem9776, carbadox metabolite, acmc-1bl7g, quinoxaline carboxylic acid
C1=CC=C2C(=C1)N=CC(=N2)C(=O)O
174.159
174.16
2-Quinoxalinecarboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.