Please login to your online account to display your discounted pricing

2-Quinoxalinecarbaldehyde, 97+%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC03504DA

 View more versions of this product

Catalog No. CC03504DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 158.16
Quantity 1g
Assay 97%
Chemical Name or Material 2-Quinoxalinecarbaldehyde
Assay Percent Range 97%
CAS 1593-08-4
Packaging Amber Glass Bottle
Molecular Formula C9H6N2O

Specifications

Chemical Name or Material 2-Quinoxalinecarbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H6N2O
Formula Weight 158.16