Please login to your online account to display your discounted pricing

2-quinolinecarboxylic acid, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Quinolinecarboxylic acid
Assay 97%
Assay Percent Range 97%
Formula Weight 173.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC04401DA

  SDS
fisher scientific company
CC04401DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.17
Quantity 1g
Assay 97%
Merck Index 15, 8162
CAS 93-10-7
Chemical Name or Material 2-Quinolinecarboxylic acid
Packaging Amber Glass Bottle
Assay Percent Range 97%