missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Pyrrolidinone hydrotribromide, 97%

$33.30 - $209.73

Chemical Identifiers

CAS 22580-55-8
Molecular Formula C4H7Br3NO-
Molecular Weight (g/mol) 324.818
MDL Number MFCD00082241
InChI Key OTYIQJFSTDUAJR-UHFFFAOYSA-N
Synonym 2-pyrrolidinone hydrotribromide, pyrrolidinone hydrotribromide, pubchem19110, pyrrolidonehydrotribromide, pyrolidone hydrotribromide, pyrolidinone hydrotribromide, pht, pyrrolidone hydrotribromide, 2?_-tribromane; pyrrolidone, pyrrolidin-2-one compound with 2l3-tribromane 1:1
PubChem CID 15287049
SMILES C1CC(=O)NC1.Br[Br-]Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0017714 View Documents Alfa Aesar™
L0017714
25g Each for $33.30
 
AAL0017722 View Documents Alfa Aesar™
L0017722
100g Each for $78.96
 
AAL0017736 View Documents Alfa Aesar™
L0017736
500g Each for $209.73
 
Description

Description

Applications
Mild, highly selective brominating agent. Ketones can be ɑ-brominated in the presence of a double bond: the low concentration of enol form consumes the bromine as it is slowly liberated

Solubility
Very sol in alcohol, ether, benzene, chloroform, carbon disulfide. Infinitely soluble in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Chemical Identifiers

22580-55-8
324.818
OTYIQJFSTDUAJR-UHFFFAOYSA-N
15287049
C4H7Br3NO-
MFCD00082241
2-pyrrolidinone hydrotribromide, pyrrolidinone hydrotribromide, pubchem19110, pyrrolidonehydrotribromide, pyrolidone hydrotribromide, pyrolidinone hydrotribromide, pht, pyrrolidone hydrotribromide, 2?_-tribromane; pyrrolidone, pyrrolidin-2-one compound with 2l3-tribromane 1:1
C1CC(=O)NC1.Br[Br-]Br

Specifications

89°C to 91°C
22580-55-8
MFCD00082241
OTYIQJFSTDUAJR-UHFFFAOYSA-N
324.818
496.05
2-Pyrrolidinone hydrotribromide
25g
C4H7Br3NO-
2-pyrrolidinone hydrotribromide, pyrrolidinone hydrotribromide, pubchem19110, pyrrolidonehydrotribromide, pyrolidone hydrotribromide, pyrolidinone hydrotribromide, pht, pyrrolidone hydrotribromide, 2?_-tribromane; pyrrolidone, pyrrolidin-2-one compound with 2l3-tribromane 1:1
C1CC(=O)NC1.Br[Br-]Br
15287049
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning